missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) iodide, 99%, ACROS Organics™

Manufacturer:  Acros Organics 198860500

 View more versions of this product

Catalog No. AC198860500



Specifications

Specifications

Lead(II) iodide
10101-63-0
I2Pb
MFCD00011163
blei ii-iodid
I[Pb]I
461
461
≥98.5%
6.1600g/mL
Glass bottle
Yellow to Orange
50g
99%
99%
PbI2
15, 5466
RQQRAHKHDFPBMC-UHFFFAOYSA-L
diiodolead
24931
Powder
100ppm max.
0.1% max.
6.16
402°C

Chemical Identifiers

10101-63-0
461
RQQRAHKHDFPBMC-UHFFFAOYSA-L
24931
I[Pb]I
I2Pb
MFCD00011163
blei ii-iodid
diiodolead
Safety and Handling

Safety and Handling

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.
Harmful if inhaled.
May damage the unborn child. Suspected of damaging fertility.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 233-256-9

TSCA : TSCA

SDS Documents