missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) iodide, 98.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 5710318

 View more versions of this product

Catalog No. AA5710318



Specifications

Specifications

Lead(II) iodide
98.5%
MFCD00011163
lead ii iodide, e2-plumbane, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, lead ii iodide, diiodo-
I[Pb]I
461.009
461
6.16g/cm3 at 20°C
Yellow
10101-63-0
I2Pb
Yellow powder
RQQRAHKHDFPBMC-UHFFFAOYSA-L
diiodolead
24931
Powder
Odorless
50g

Chemical Identifiers

10101-63-0
461.009
RQQRAHKHDFPBMC-UHFFFAOYSA-L
24931
I[Pb]I
I2Pb
MFCD00011163
lead ii iodide, e2-plumbane, lead diiodide, plumbous iodide, lead iodide pbi2, pbi2, plumbum jodatum, blei ii-iodid, hsdb 636, lead ii iodide, diiodo-
diiodolead
Safety and Handling

Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 233-256-9

SDS Documents