Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) hexafluorosilicate dihydrate

Chemical Identifiers

Chemical Name or Material Lead(II) hexafluorosilicate dihydrate
CAS 1310-03-8
Linear Formula PbSiF6·2H2O
Formula Weight 385.26 (349.26 Anhydrous)
MDL Number MFCD00049509
UN Number UN2291
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3934709

  SDS
alfa aesar™
3934709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3934718

  SDS
alfa aesar™
3934718
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 385.26 (349.26 Anhydrous)
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
UN Number UN2291
Health Hazard 1 Danger
TSCA TSCA
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Lead(II) hexafluorosilicate dihydrate
RTECS Number VV8450000
CAS 1310-03-8
Linear Formula PbSiF6·2H2O
MDL Number MFCD00049509
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Physical Form Powder
Odor Odorless
Quantity 10g
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.