missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) fluoride, 99% min

Manufacturer:  Alfa Aesar™ 8828218*

 View more versions of this product

Catalog No. AA8828218



Specifications

Specifications

Lead (II) fluoride
F2Pb
UN2291
lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure
F[Pb]F
245.197
245.2
≥99%
Odorless
855°C
7783-46-2
MFCD00011162
14,5408
FPHIOHCCQGUGKU-UHFFFAOYSA-L
difluorolead
24549
Powder
8.445g/mL
1290°C
50g

Chemical Identifiers

7783-46-2
245.197
FPHIOHCCQGUGKU-UHFFFAOYSA-L
24549
F[Pb]F
F2Pb
MFCD00011162
lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure
difluorolead
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

TSCA : TSCA

SDS Documents