Learn More
Lead(II) fluoride, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 08828230
Description
Lead(II) fluoride is a specialty product for proteomics research. It is used in low melting glasses, in glass coatings to reflect infrared rays, in phosphors for television-tube screens and as a catalyst for the manufacture of picoline.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1290°C | |
855°C | |
Powder | |
F2Pb | |
UN2291 | |
lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure | |
FPHIOHCCQGUGKU-UHFFFAOYSA-L | |
difluorolead | |
24549 | |
≥99% | |
8.445 g/mL |
Lead (II) fluoride | |
7783-46-2 | |
250 g | |
MFCD00011162 | |
14,5408 | |
Sparingly soluble in water(0.64g/L at 20°C). Soluble in nitric acid. Insoluble in acetone and ammonia. | |
[F-].[F-].[Pb++] | |
245.20 | |
245.2 | |
Odorless |
Chemical Identifiers
7783-46-2 | |
245.20 | |
FPHIOHCCQGUGKU-UHFFFAOYSA-L | |
24549 | |
[F-].[F-].[Pb++] |
F2Pb | |
MFCD00011162 | |
lead fluoride, lead ii fluoride, lead difluoride, lead fluoride pbf2, plumbous fluoride, lead +2 fluoride, difluoroplumbane, plomb fluorure french, pbf2, plomb fluorure | |
difluorolead |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
EINECSNumber : 231-998-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only