Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) cyclohexanebutyrate, 94%

Chemical Identifiers

Chemical Name or Material Lead(II) cyclohexanebutyrate
CAS 62637-99-4
Molecular Formula C20H34O4Pb
Formula Weight 545.68
MDL Number MFCD00036403
UN Number UN2291
Synonym Cyclohexanebutyric acid lead(II) salt; 4-Cyclohexylbutyric acid lead(II) salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1728006

  SDS
alfa aesar™
A1728006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1728014

  SDS
alfa aesar™
A1728014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1728022

  SDS
alfa aesar™
A1728022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 545.68
Chemical Name or Material Lead(II) cyclohexanebutyrate
TSCA TSCA
Quantity 5g
Assay 94%
Molecular Formula C20H34O4Pb
MDL Number MFCD00036403
CAS 62637-99-4
UN Number UN2291
Synonym Cyclohexanebutyric acid lead(II) salt; 4-Cyclohexylbutyric acid lead(II) salt
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.