missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) cyanurate monohydrate

Manufacturer:  Alfa Aesar™ 2255606

 View more versions of this product

Catalog No. AA2255606



Specifications

Specifications

Lead(II) cyanurate monohydrate
53846-29-0
MFCD00064816
lead cyanurate, lead ii cyanurate monohydrate, lead 2+ ;1,3,5-triazine-2,4,6-triolate;trihydrate
C1(=NC(=NC(=N1)[O-])[O-])[O-].C1(=NC(=NC(=N1)[O-])[O-])[O-].O.O.O.[Pb+2].[Pb+2].[Pb+2]
927.747
891.69 (873.67 Anhydrous)
5g
C6H6N6O9Pb3
UN3467
DPLKIANDZYQWKF-UHFFFAOYSA-H
lead(2+);1,3,5-triazine-2,4,6-triolate;trihydrate
90473286
Powder

Chemical Identifiers

53846-29-0
927.747
DPLKIANDZYQWKF-UHFFFAOYSA-H
90473286
C1(=NC(=NC(=N1)[O-])[O-])[O-].C1(=NC(=NC(=N1)[O-])[O-])[O-].O.O.O.[Pb+2].[Pb+2].[Pb+2]
C6H6N6O9Pb3
MFCD00064816
lead cyanurate, lead ii cyanurate monohydrate, lead 2+ ;1,3,5-triazine-2,4,6-triolate;trihydrate
lead(2+);1,3,5-triazine-2,4,6-triolate;trihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.

SDS Documents