Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) cyanurate monohydrate

Chemical Identifiers

Chemical Name or Material Lead(II) cyanurate monohydrate
CAS 53846-29-0
Formula Weight 891.69 (873.67 Anhydrous)
MDL Number MFCD00064816
UN Number UN3467
Synonym Cyanuric acid lead(II) salt monohydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2255606

  SDS
alfa aesar™
2255606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2255614

  SDS
alfa aesar™
2255614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 891.69 (873.67 Anhydrous)
Chemical Name or Material Lead(II) cyanurate monohydrate
Quantity 5g
Physical Form Powder
MDL Number MFCD00064816
CAS 53846-29-0
UN Number UN3467
Synonym Cyanuric acid lead(II) salt monohydrate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOMETALLIC COMPOUND, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.