Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) citrate trihydrate, 99%

Chemical Identifiers

Chemical Name or Material Lead (II) citrate trihydrate
CAS 6107-83-1
Molecular Formula C12H10O14Pb3·3H2O
Formula Weight 1053.82 (999.77 Anhydrous)
MDL Number MFCD00150456
UN Number UN2291
Synonym Citric acid lead(II) salt trihydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1070122

 
alfa aesar™
A1070122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1070136

 
alfa aesar™
A1070136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1053.82 (999.77 Anhydrous)
Chemical Name or Material Lead (II) citrate trihydrate
TSCA TSCA
Assay 99%
Molecular Formula C12H10O14Pb3·3H2O
MDL Number MFCD00150456
CAS 6107-83-1
UN Number UN2291
Synonym Citric acid lead(II) salt trihydrate
Solubility Information Soluble in cold water
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 100g
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.