Please login to your online account to display your discounted pricing

Lead(II) chromate, ACS, 98%, Alfa Aesar™

Maximum level of impurities: Soluble matter 0.15%, Carbon compounds (as C) 0.01%

Chemical Identifiers

Chemical Name or Material Lead(II) chromate
Name Note For ACS analysis
CAS 7758-97-6
Purity Grade ACS Grade
Physical Form Powder
Assay Percent Range 98%
Molecular Formula CrO4Pb
Formula Weight 323.19
MDL Number MFCD00011158
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1412522

  SDS
alfa aesar
1412522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1412536

  SDS
alfa aesar
1412536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Lead(II) chromate
Name Note For ACS analysis
CAS 7758-97-6
Health Hazard 1 H350-H360-H373-H400-H410: May cause cancer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOT Information DOT Class: 9, Packaging Group: III
EINECS Number 231-846-0
Purity Grade ACS Grade
Physical Form Powder
Assay Percent Range 98%
Molecular Formula CrO4Pb
Appearance Yellow powder
Formula Weight 323.19
MDL Number MFCD00011158
Density 6.3g/cm3 at 20°C
Odor Odorless
Color Yellow
Quantity 100g
Safety and Handling

H350-H360-H373-H400-H410: May cause cancer. May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.