Please login to your online account to display your discounted pricing

Lead(II) chloride, ultra dry, 99.999% (metals basis), Alfa Aesar™

$25.39 - $25.39

Chemical Identifiers

Chemical Name or Material Lead(II) chloride, ultra dry
CAS 7758-95-4
Physical Form Powder
Assay Percent Range 99.999% (Metal basis)
Molecular Formula PbCl2
Formula Weight 278.1
MDL Number MFCD00011157
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4284104

 
alfa aesar
4284104
2g Each for $25.39

AA4284118

 
alfa aesar
4284118
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 278.1
Physical Form Powder
Quantity 2g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Lead(II) chloride, ultra dry
EINECS Number 231-845-5
Odor Odorless
Assay Percent Range 99.999% (Metal basis)
Molecular Formula PbCl2
Appearance White powder
MDL Number MFCD00011157
CAS 7758-95-4
Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
Density 5.85g/cm3 at 20°C
Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.