Learn More
Lead(II) chloride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193312500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
950.0°C | |
501.0°C | |
White | |
250 g | |
Cl2Pb | |
MFCD00011157 | |
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry | |
HWSZZLVAJGOAAY-UHFFFAOYSA-L | |
λ²-lead(2+) dichloride | |
24459 | |
99% |
Lead(II) chloride | |
7758-95-4 | |
Crystalline Powder | |
98.5% min. (Complexometry) | |
PbCl2 | |
15, 5459 | |
Solubility in water: 0.99g/L (20°C). Other solubilities: readly soluble in solution of nh4cl and nh4no3, soluble in alkalis | |
[Cl-].[Cl-].[Pb++] | |
278.10 | |
278.11 | |
Plastic bottle |
Chemical Identifiers
7758-95-4 | |
278.10 | |
HWSZZLVAJGOAAY-UHFFFAOYSA-L | |
24459 |
Cl2Pb | |
MFCD00011157 | |
lead dichloride, lead ii chloride, lead chloride, plumbous chloride, lead 2+ chloride, lead chloride pbcl2, pbcl2, ccris 7565, lead ii chloride, ultra dry | |
[Cl-].[Cl-].[Pb++] |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes damage to organs through prolonged or repeated exposure if inhaled.
Very toxic to aquatic life with long lasting effects.
May damage the unborn child.
Su
GHS P Statement
Use personal protective equipment as required.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON C
GHS Signal Word: Danger
EINECSNumber : 231-845-5
RUO – Research Use Only