missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) carbonate, White powder, Puratronic™, 99.999% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1072114

 View more versions of this product

Catalog No. AA1072114



Specifications

Specifications

Lead(II) carbonate
99.999% (Metals basis)
MFCD00064631
MFEVGQHCNVXMER-UHFFFAOYSA-L
lead(2+);carbonate
11727
Powder
White
598-63-0
CO3Pb
carbonic acid, lead 2+ salt 1:1
C(=O)([O-])[O-].[Pb+2]
267.208
267.2
Odorless
25g

Chemical Identifiers

598-63-0
267.208
MFEVGQHCNVXMER-UHFFFAOYSA-L
11727
C(=O)([O-])[O-].[Pb+2]
CO3Pb
MFCD00064631
carbonic acid, lead 2+ salt 1:1
lead(2+);carbonate
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 209-943-4

Documents