Please login to your online account to display your discounted pricing

Lead(II) carbonate, ACS, Alfa Aesar™

$254.65 - $254.65

Chemical Identifiers

Chemical Name or Material Lead(II) carbonate
CAS 598-63-0
Purity Grade ACS Grade
Molecular Formula PbCO3
Formula Weight 267.2
MDL Number MFCD00064631
UN Number UN2291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4011936

  SDS
alfa aesar
4011936
Each for $254.65
Description & Specifications

Specifications

Chemical Name or Material Lead(II) carbonate
CAS 598-63-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H373-H302-H332;
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.
TSCA TSCA
Purity Grade ACS Grade
Physical Form Powder
Molecular Formula PbCO3
Formula Weight 267.2
MDL Number MFCD00064631
UN Number UN2291
Odor Odorless
Quantity 500g
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332;
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.