Please login to your online account to display your discounted pricing

Lead(II) bromide, White powder, 99.999% (Metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Lead(II) bromide
CAS 10031-22-8
Physical Form Powder
Assay Percent Range 99.999% (Metals basis)
Molecular Formula Br2Pb
Formula Weight 367.01
MDL Number MFCD00011156
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3570303

 
alfa aesar
3570303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3570306

 
alfa aesar
3570306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3570314

 
alfa aesar
3570314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 367.01
Quantity 1g
Color White
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Lead(II) bromide
EINECS Number 233-084-4
Odor Odorless
Assay Percent Range 99.999% (Metals basis)
Molecular Formula Br2Pb
MDL Number MFCD00011156
CAS 10031-22-8
Health Hazard 1 H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.
Safety and Handling

H360-H373-H400-H410-H302-H332: May damage fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled.