missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) bromide, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1072014

 View more versions of this product

Catalog No. AA1072014



Specifications

Specifications

Lead(II) bromide
(metals basis)
PbBr2
UN2291
acmc-1bq2x
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
dibromolead
24831
Powder
Puratronic™
Odorless
White
25g
10031-22-8
Br2Pb
MFCD00011156
14,5403
Soluble in water; Insoluble in alcohol
Br[Pb]Br
367.008
367.01
99.999%
6.66g/mL
916°C
373°C

Chemical Identifiers

10031-22-8
367.008
ZASWJUOMEGBQCQ-UHFFFAOYSA-L
24831
Br[Pb]Br
Br2Pb
MFCD00011156
acmc-1bq2x
dibromolead
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD COMPOUNDS, SOLUBLE, N.O.S.

TSCA : TSCA

Documents