missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead(II) acetate trihydrate, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 1071930

 View more versions of this product

Catalog No. AA1071930



Specifications

Specifications

Lead(II) acetate trihydrate
(metals basis)
MFCD00150023
3730298
acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate, bis acetato trihydroxytrilead, bleiazetat german, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, lead acetate trihydrate, lead diacetate trihydrate, unii-rx077p88ry
MCEUZMYFCCOOQO-UHFFFAOYSA-L
lead(2+);diacetate;trihydrate
22456
379.33 (325.28 Anhydrous)
99.995%
2.55 g/mL
250g
6080-56-4
C4H12O7Pb
UN1616
14,5397
Freely soluble in water,alcohol and glycerol.
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
379.333
CHEBI:33112
Crystalline
Puratronic™
75°C

Chemical Identifiers

6080-56-4
379.333
MCEUZMYFCCOOQO-UHFFFAOYSA-L
22456
lead(2+);diacetate;trihydrate
C4H12O7Pb
MFCD00150023
acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate, bis acetato trihydroxytrilead, bleiazetat german, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, lead acetate trihydrate, lead diacetate trihydrate, unii-rx077p88ry
CHEBI:33112
CC(=O)[O-].CC(=O)[O-].O.O.O.[Pb+2]
Safety and Handling

Safety and Handling

GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c

H360D-H361f-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE

EINECSNumber : 206-104-4

RTECSNumber : OF8050000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents