Learn More
Lead(II) acetate trihydrate, ACS, 99.0-103.0%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 014242A3
Description
Lead(II) acetate trihydrate is used as a mordant in textile printing and dyeing, drier in paints, varnishes and as a water repellant. It is also used in cosmetics and to prepare other lead compounds. It serves as an ingredient in progressive types of hair coloring dyes. Further, it is used in the detection of poisonous gas hydrogen sulfide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Lead(II) acetate trihydrate | |
6080-56-4 | |
2 kg | |
MFCD00150023 | |
3730298 | |
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
λ²-lead(2+) diacetate trihydrate | |
22456 | |
379.33 (325.28 Anhydrous) | |
ACS |
75°C | |
Granules | |
C4H12O7Pb | |
UN1616 | |
14,5397 | |
Freely soluble in water,alcohol and glycerol. | |
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O | |
379.30 | |
CHEBI:33112 | |
99.0 to 103.0% | |
2.55 g/mL |
Chemical Identifiers
6080-56-4 | |
379.30 | |
MCEUZMYFCCOOQO-UHFFFAOYSA-L | |
22456 | |
O.O.O.[Pb++].CC([O-])=O.CC([O-])=O |
C4H12O7Pb | |
MFCD00150023 | |
lead acetate trihydrate, lead diacetate trihydrate, bleiazetat german, bis acetato trihydroxytrilead, unii-rx077p88ry, ccris 9031, lead acetate ii , trihydrate, lead acetate jan, acetic acid, lead +2 salt trihydrate, acetic acid, lead 2+ salt, trihydrate | |
CHEBI:33112 |
Safety and Handling
GHS H Statement
H360-H373
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H360D-H361f-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: LEAD ACETATE
EINECSNumber : 206-104-4
RTECSNumber : OF8050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only