Please login to your online account to display your discounted pricing

Lead(II) acetate trihydrate, 99+%, for analysis, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  201750250

 View more versions of this product

Catalog No. AC201750250

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Formula Weight 379.33
Quantity 25g
Color White
Merck Index 15, 5452
Purity Grade For analysis
Assay 99.0 to 103.0%
RTECS Number OF8050000
Packaging Plastic bottle
Potassium (K) 50ppm max.
Sodium (Na) 50ppm max.
Name Note p.a.
Assay Percent Range 99+%
Molecular Formula C4H6O4Pb·3H2O
Linear Formula (CH3CO2)2Pb·3H2O
CAS 6080-56-4
Health Hazard 1 Danger
Chemical Name or Material Lead(II) acetate trihydrate
Iron (Fe) 10ppm max.
Nitrate 50ppm max.
MDL Number MFCD00150023
Synonym Acetic acid, lead salt trihydrate
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.
Calcium (Ca) 50ppm max.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chloride 5ppm max.
Copper (Cu) 10ppm max.

Specifications

Chemical Name or Material Lead(II) acetate trihydrate
Name Note p.a.
CAS 6080-56-4
Purity Grade For analysis
Assay 99.0 to 103.0%
Assay Percent Range 99+%
Molecular Formula C4H6O4Pb·3H2O
Linear Formula (CH3CO2)2Pb·3H2O
Formula Weight 379.33
MDL Number MFCD00150023
Synonym Acetic acid, lead salt trihydrate
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents