missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead(II) acetate trihydrate, 99+%, ACS reagent, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 423845000

 View more versions of this product

Catalog No. AC423845000


 

Specifications Description & Specifications

Specifications

Lead(II) acetate trihydrate
ACS Reagent Grade
99.0 to 103.0%
C4H6O4Pb·3H2O
379.33
01,532; 02,233; 04,276
Acetic acid, lead salt trihydrate
5ppm max.
13.1
10ppm max.
Plastic bottle
100ppm max.
White
6080-56-4
Crystals or Crystalline Powder
99+%
(CH3CO2)2Pb·3H2O
MFCD00150023
15, 5452
50ppm max.
20ppm max.
0.01% max.
0.005% max. (as NO3)
50ppm max.
Decomposes information: >100°C
500g

Chemical Identifiers

6080-56-4
C4H6O4Pb·3H2O
379.33
Acetic acid, lead salt trihydrate
99.0 to 103.0%
99+%
(CH3CO2)2Pb·3H2O
MFCD00150023
Lead(II) acetate trihydrate
ACS Reagent Grade

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

Emergency Overview
Contains a known or suspected carcinogen. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of serious damage to health by prolonged exposure. Danger of cumulative effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment.
NFPA
Health:2 Flammability:0 Instability:0

Signal Word:DANGER!

RTECSNumber : OF8050000