Please login to your online account to display your discounted pricing

Lead(II) acetate trihydrate, 99.999%, (trace metal basis), extra pure, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  317230250

 View more versions of this product

Catalog No. AC317230250

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Trace Metal Basis
Physical Form Crystals or Crystalline Powder
Formula Weight 379.33
Quantity 25g
Color White
Merck Index 15, 5452
RTECS Number OF8050000
Fieser 01,532; 02,233; 04,276
Chemical Name or Material Lead(II) acetate trihydrate
Name Note 99.999%
Packaging Glass bottle
Assay Percent Range 99.999%
Molecular Formula C4H6O4Pb·3H2O
Linear Formula (CH3CO2)2Pb·3H2O
MDL Number MFCD00150023
CAS 6080-56-4
Total Trace Metal Impurities 0.001% max.
Synonym Acetic acid, lead salt trihydrate
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Lead(II) acetate trihydrate
Name Note 99.999%
CAS 6080-56-4
Purity Grade Trace Metal Basis
Assay Percent Range 99.999%
Molecular Formula C4H6O4Pb·3H2O
Linear Formula (CH3CO2)2Pb·3H2O
Formula Weight 379.33
MDL Number MFCD00150023
Synonym Acetic acid, lead salt trihydrate
Accessories

Danger

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure if swallowed.
May damage the unborn child. Suspected of damaging fertility.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents