Learn More
Lead(II) 2-ethylhexanoate, Thermo Scientific Chemicals
$104.11 - $209.60
Chemical Identifiers
CAS | 301-08-6 |
---|---|
Molecular Formula | C16H30O4Pb |
Molecular Weight (g/mol) | 493.612 |
MDL Number | MFCD00014003 |
InChI Key | RUCPTXWJYHGABR-UHFFFAOYSA-L |
Synonym | lead bis 2-ethylhexanoate, lead ii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lead salt, hexanoic acid, 2-ethyl-, lead salt 1:?, lead tallate 2-ethylhexanoate, lead 2+ bis 2-ethylhexanoate, cis-9-octadecenoamide, plumbous 2-ethylhexanoate, acmc-1aj1i, 2-ethylhexanoate; lead 2+ |
PubChem CID | 160451 |
IUPAC Name | 2-ethylhexanoate;lead(2+) |
SMILES | CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Pb+2] |
Description
Films of lead (II) 2-ethylhexanote can be cast on silicon solutions by spin coating. Lead (II) 2-ethylhexanote is a common solvent in metal-organic deposition (MOD) process. Intermetallic PtPb nanoparticles have been synthesized by the chemical reduction of dimethyl(1,5-cyclooctadiene)platinum and lead(II)2-ethylhexanoate by sodium naphthalide in THF or diglyme. The precursors used for the photochemical production of PZT or PLZT films were lead(II) 2-ethylhexanoate, lanthanum(III) 2-ethylhexanoate, zirconyl(IV) 2-ethylhexa- noate, and titanium(IV) isopropoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
301-08-6 | |
493.612 | |
RUCPTXWJYHGABR-UHFFFAOYSA-L | |
160451 | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Pb+2] |
C16H30O4Pb | |
MFCD00014003 | |
lead bis 2-ethylhexanoate, lead ii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lead salt, hexanoic acid, 2-ethyl-, lead salt 1:?, lead tallate 2-ethylhexanoate, lead 2+ bis 2-ethylhexanoate, cis-9-octadecenoamide, plumbous 2-ethylhexanoate, acmc-1aj1i, 2-ethylhexanoate; lead 2+ | |
2-ethylhexanoate;lead(2+) |
Specifications
301-08-6 | |
Odorless | |
MFCD00014003 | |
UN3082 | |
Insoluble in water. | |
CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Pb+2] | |
493.612 | |
493.61 | |
Lead(II) 2-ethylhexanoate |
1.56 g/mL | |
C16H30O4Pb | |
50 g | |
lead bis 2-ethylhexanoate, lead ii 2-ethylhexanoate, hexanoic acid, 2-ethyl-, lead salt, hexanoic acid, 2-ethyl-, lead salt 1:?, lead tallate 2-ethylhexanoate, lead 2+ bis 2-ethylhexanoate, cis-9-octadecenoamide, plumbous 2-ethylhexanoate, acmc-1aj1i, 2-ethylhexanoate; lead 2+ | |
RUCPTXWJYHGABR-UHFFFAOYSA-L | |
2-ethylhexanoate;lead(2+) | |
160451 | |
Viscous Liquid |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P501c
H302+H332-H350-H360Df-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 206-107-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only