missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lead ICP Standard, 1000 ppm Pb in 3% HNO3, Ricca Chemical
$265.44 - $265.44
Chemical Identifiers
CAS | 7439-92-1 |
---|---|
Molecular Formula | Pb |
Molecular Weight (g/mol) | 207.20 |
MDL Number | MFCD00134050 |
InChI Key | WABPQHHGFIMREM-UHFFFAOYSA-N |
PubChem CID | 5352425 |
ChEBI | CHEBI:27889 |
IUPAC Name | lead |
SMILES | [Pb] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
PPB1KN-100
|
Ricca Chemical Company
PPB1KN100 |
100 mL | Natural LDPE Bottle |
Each for $265.44
|
|
||||
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
CHEBI:27889 | |
[Pb] |
Pb | |
MFCD00134050 | |
5352425 | |
lead |
Specifications
7439-92-1,7697-37-2,7439-92-1,7732-18-5 | |
100 mL | |
97.0,5.09,0.1 | |
1,000ppm Pb | |
MFCD00134050 | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
Liquid |
Natural LDPE Bottle | |
93.0,4.89,0.09 | |
Colorless | |
Pb | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
1 mL = 1 mg Pb | |
Lead ICP Standard |