Please login to your online account to display your discounted pricing

Alfa Aesar™ Lead granules, approximately 3mm (0.12 in.), 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Lead granules
CAS 7439-92-1
Name Note Approximately 3mm (0.12 in.)
MDL Number MFCD00134050
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3968518

  SDS
alfa aesar™
3968518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3968530

  SDS
alfa aesar™
3968530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note Approximately 3mm (0.12 in.)
CAS 7439-92-1
Color Gray
RTECS Number OF7525000
Physical Form Granules
Quantity 50g
MDL Number MFCD00134050
Chemical Name or Material Lead granules
Health Hazard 1 Danger
Assay 99.9%
Assay Percent Notes (metals basis)
Health Hazard 2 GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Odor Odorless
Safety and Handling

Danger

GHS P Statement
P260-P261-P281-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use personal protective equipment as required.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.