missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead Foil, 1.0mm (0.04 in.) thick, Puratronic™, 99.998% (Metals basis), Alfa Aesar™

Form: Approx.7.1g/25 x 25mm

Manufacturer:  Thermo Fisher Scientific 12048FF

 View more versions of this product

Catalog No. AA12048FF



Specifications

Specifications

Lead
7439-92-1
99.998% (Metals basis)
MFCD00134050
WABPQHHGFIMREM-UHFFFAOYSA-N
lead
207.2
CHEBI:27889
Gray
25 x 25mm
Gray foil
Pb
blei, element, flake, glover, lead, elemental, metal, omaha & grant, plomo, plumbum, rough bullion
[Pb]
Odorless
5352425
Foil, 1.0mm (0.04 in.) thick

Chemical Identifiers

7439-92-1
207.2
WABPQHHGFIMREM-UHFFFAOYSA-N
5352425
lead
Pb
MFCD00134050
blei, element, flake, glover, lead, elemental, metal, omaha & grant, plomo, plumbum, rough bullion
CHEBI:27889
[Pb]
Safety and Handling

Safety and Handling

Avoid exposure - obtain special instructions before use. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c

H350-H360FD-H362-H372

EINECSNumber : 231-100-4

RTECSNumber : OF7525000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents