Learn More
Lead foil, 0.5mm (0.02in) thick, Puratronic, 99.99% (metalsbasis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012049FI
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Gray | |
0.5mm (0.02 in.) thick | |
(metals basis) | |
MFCD00134050 | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
[Pb] | |
207.20 | |
CHEBI:27889 | |
Puratronic™ | |
≈3.5g/25x25mm |
Lead foil | |
7439-92-1 | |
Pb | |
50 x 50 mm | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
lead | |
5352425 | |
99.998% | |
Odorless | |
Foil |
Chemical Identifiers
7439-92-1 | |
207.20 | |
WABPQHHGFIMREM-UHFFFAOYSA-N | |
5352425 | |
lead |
Pb | |
MFCD00134050 | |
plumbum, metal, blei, lead, elemental, rough bullion, plomo, glover, element, flake, omaha & grant | |
CHEBI:27889 | |
[Pb] |
Safety and Handling
GHS H Statement
H360-H373-H302-H332
May damage fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P260-P263-P264b-P270-P281-P308+P313-P501c
H350-H360FD-H362-H372
EINECSNumber : 231-100-4
RTECSNumber : OF7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only