Please login to your online account to display your discounted pricing

Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate), 99+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  347800250

 View more versions of this product

Catalog No. AC347800250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder
Formula Weight 573.74
Quantity 25g
Color White
Melting Point 128° to 131°C
Infrared Spectrum Authentic
Lead (Pb) 35.75% min.
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate)
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C22 H38 O4 Pb
MDL Number MFCD00070468
CAS 21319-43-7
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertil

Specifications

Chemical Name or Material Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate)
Name Note 99+%
CAS 21319-43-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C22 H38 O4 Pb
Formula Weight 573.74
MDL Number MFCD00070468
Accessories

GHS Signal Word: Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertil

SDS & Safety Documents