Please login to your online account to display your discounted pricing

Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate), 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate)
Name Note 99+%
CAS 21319-43-7
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C22 H38 O4 Pb
Formula Weight 573.74
MDL Number MFCD00070468
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC347800050

  SDS
thermo fisher scientific
347800050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC347800250

  SDS
thermo fisher scientific
347800250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥99.0%
Physical Form Powder
Formula Weight 573.74
Quantity 5g
Color White
Melting Point 128° to 131°C
Chemical Name or Material Lead bis(2,2,6,6-tetramethyl-3,5-heptanedionate)
Assay Percent Range 99+%
MDL Number MFCD00070468
CAS 21319-43-7
Infrared Spectrum Authentic
Lead (Pb) 35.75% min.
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertil
Health Hazard 3 GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 99+%
Molecular Formula C22 H38 O4 Pb
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertil