Please login to your online account to display your discounted pricing

Lead Acetate, Trihydrate, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$315.28 - $315.28

Chemical Identifiers

Chemical Name or Material Lead Acetate
CAS 6080-56-4
Name Note Trihydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula (CH3COO)2Pb·3H2O
Formula Weight 379.33
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003355

 
avantor performance
227101
Poly Bottle 500g Each for $315.28
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 379.33
Chemical Name or Material Lead Acetate
Molecular Formula (CH3COO)2Pb·3H2O
CAS 6080-56-4
Name Note Trihydrate
Packaging Poly Bottle
Physical Form Granular
Specific Gravity 1L = 2.55kg
Quantity 500g