Please login to your online account to display your discounted pricing

Lead Acetate Trihydrate (Certified ACS), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  L33500

 View more versions of this product

Catalog No. L33-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified ACS Grade
Formula Weight 379.3
Melting Point 75°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Assay 99 to 103%
Physical Form Solid
Molecular Formula C4H6O4Pb · 3 H2O
pH 5.5 to 6.5
Decomposition Information Decomposes information: >100°C
Color White
UN Number 1616
Identification Pass Test
Copper (Cu) 0.002% max.
Nitrate and Nitrite 0.005% max.
Assay Percent Range 99 to 103 %
Chloride 5ppm max.
Health Hazard 3 Emergency Overview
Contains a known or suspected carcinogen. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of serious damage to health by prolonged exposure. Danger of cumulative effects. Very toxic to aquatic organisms
Linear Formula Pb(C2H3O2)2·3H2O
Chemical Name or Material Lead Acetate Trihydrate
CAS 6080-56-4
MDL Number MFCD00150023
Potassium (K) 0.005% max.
Packaging Glass Bottle
Synonym Lead (II) Acetate Trihydrate
Calcium (Ca) 0.005% max.
Iron (Fe) 0.001% max.
Sodium (Na) 0.01% max.
Insoluble Matter 0.01% max.
Health Hazard 2 WARNING!
Health Hazard 1 Signal Word:DANGER!
ChemAlert Storage Symbol Gray

Specifications

Chemical Name or Material Lead Acetate Trihydrate
CAS 6080-56-4
Purity Grade Certified ACS Grade
Assay 99 to 103%
Assay Percent Range 99 to 103 %
Molecular Formula C4H6O4Pb · 3 H2O
Linear Formula Pb(C2H3O2)2·3H2O
Formula Weight 379.3
MDL Number MFCD00150023
Synonym Lead (II) Acetate Trihydrate
Accessories
 

Signal Word:DANGER!

Emergency Overview
Contains a known or suspected carcinogen. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of serious damage to health by prolonged exposure. Danger of cumulative effects. Very toxic to aquatic organisms

WARNING!

SDS & Safety Documents