Please login to your online account to display your discounted pricing

Lead Acetate Trihydrate (Certified ACS), Fisher Chemical

$206.69 - $222.31

Chemical Identifiers

Chemical Name or Material Lead Acetate Trihydrate
CAS 6080-56-4
Purity Grade Certified ACS Grade
Assay 99 to 103%
Assay Percent Range 99 to 103 %
Molecular Formula C4H6O4Pb · 3 H2O
Linear Formula Pb(C2H3O2)2·3H2O
Formula Weight 379.3
MDL Number MFCD00150023
Synonym Lead (II) Acetate Trihydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

L33-500

  SDS
fisher scientific company
L33500
Glass Bottle 500g Each for $222.31
Case of 6 Each for $1,333.86

L33-250

  SDS
fisher scientific company
L33250
Glass Bottle 250g Each for $206.69
Case of 6 Each for $1,240.14
Description & Specifications

Specifications

Purity Grade Certified ACS Grade
Formula Weight 379.3
Melting Point 75°C
Quantity 500g
DOT Information DOT Class 6.1, : Poison
Physical Form Solid
Assay 99 to 103%
Molecular Formula C4H6O4Pb · 3 H2O
Viscosity [NULL]
Decomposition Information Decomposes information: >100°C
Color White
pH 5.5 to 6.5
UN Number 1616
Identification Pass Test
Nitrate and Nitrite 0.005% max.
Copper (Cu) 0.002% max.
Chemical Name or Material Lead Acetate Trihydrate
Insoluble Matter 0.01% max.
Iron (Fe) 0.001% max.
Packaging Glass Bottle
Potassium (K) 0.005% max.
Assay Percent Range 99 to 103 %
Sodium (Na) 0.01% max.
Linear Formula Pb(C2H3O2)2·3H2O
MDL Number MFCD00150023
CAS 6080-56-4
ChemAlert Storage Symbol Gray
Synonym Lead (II) Acetate Trihydrate
Health Hazard 1 Signal Word:DANGER!
Health Hazard 2 WARNING!
Calcium (Ca) 0.005% max.
Health Hazard 3 Emergency Overview
Contains a known or suspected carcinogen. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of serious damage to health by prolonged exposure. Danger of cumulative effects. Very toxic to aquatic organisms
Chloride 5ppm max.
Safety and Handling
 

Signal Word:DANGER!

Emergency Overview
Contains a known or suspected carcinogen. May cause central nervous system effects. May cause harm to the unborn child. Possible risk of impaired fertility. Danger of serious damage to health by prolonged exposure. Danger of cumulative effects. Very toxic to aquatic organisms

WARNING!