Please login to your online account to display your discounted pricing

Lead Acetate, Trihydrate, ACS Grade, 99.0 to 103.0%, LabChem™

$175.91 - $175.91

Chemical Identifiers

Chemical Name or Material Lead Acetate
CAS 6080-56-4
CAS Max % 100
Name Note Trihydrate
Purity Grade ACS Grade
Linear Formula Pb(COOCH3)2·3H2O
Formula Weight 379.33
UN Number UN1616
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC160801

 
lab chem
LC160801
500g Each for $175.91
Description & Specifications

Specifications

Formula Weight 379.33
Chemical Name or Material Lead Acetate
EINECS Number 206-104-4
Identification Passes Test
Name Note Trihydrate
Packaging Amber Glass
Purity Grade ACS Grade
Physical Form Crystals
Assay 99.0 to 103.0%
Solubility Soluble in water
Specific Gravity 2.55g/cm3
Storage Information Room Temperature
Linear Formula Pb(COOCH3)2·3H2O
CAS 6080-56-4
UN Number UN1616
Decomposition Information Lead oxide; Carbon monoxide; Carbon dioxide
Color White
Melting Point 75°C
Quantity 500g
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
May damage fertility or the unborn child.
May cause damage to organs (kidneys, liver, blood, brain) through prolonged or repeated exposure (oral).
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Get medical advice and attention if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2.55g/cm3
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe dust.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Get medical advice and attention if you feel unwell.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May damage fertility or the unborn child.
May cause damage to organs (kidneys, liver, blood, brain) through prolonged or repeated exposure (oral).
Very toxic to aquatic life with long lasting effects.