missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lead, +99%, granular, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 222621000

 View more versions of this product

Catalog No. AC222621000


 

Specifications Description & Specifications

Specifications

≥99.0%
Granules
Gray
15, 5451
Pb
7439-92-1
MFCD00134050
Plastic bottle
207.19
100g
02,233
Pb
99+%
Lead
p.a.

Chemical Identifiers

7439-92-1
Pb
207.19
Lead, p.a.
99+%
Pb
MFCD00134050
≥99.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause damage to organs through prolonged or repeated exposure.
May damage the unborn child. Suspected of damaging fertility.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Use personal protective equipment as required.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 231-100-4

RTECSNumber : OF7525000

TSCA : TSCA