Please login to your online account to display your discounted pricing

Lead, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$111.08 - $133.51

Chemical Identifiers

Chemical Name or Material Lead, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Pb
Formula Weight 207.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003186

 
avantor performance
576504
Poly Bottle 100mL Each for $133.51

02003187

 
avantor performance
645504
Narrow Mouth Poly Bottle 150mL Each for $111.08
Description & Specifications

Specifications

Formula Weight 207.2
Chemical Name or Material Lead, 1000μg/mL (0.10% w/v)
Quantity 100mL
Specific Gravity 1L = 1.00kg
Molecular Formula Pb
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Poly Bottle