Please login to your online account to display your discounted pricing

Lead, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™


Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

Quantity 150mL
Packaging Narrow Mouth Poly Bottle

Manufacturer: avantor performance  645504

 View more versions of this product

Catalog No. 02003187

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 207.2
Chemical Name or Material Lead, 1000μg/mL (0.10% w/v)
Molecular Formula Pb
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Narrow Mouth Poly Bottle
Quantity 150mL
Specific Gravity 1L = 1.00kg

Specifications

Chemical Name or Material Lead, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Pb
Formula Weight 207.2