Please login to your online account to display your discounted pricing

LE 135, Tocris

Selective RARβ antagonist

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material LE 135
Assay Percent Range >97%
Molecular Formula C29H30N2O2
Formula Weight 438.56
Synonym 4-(7,8,9,10-Tetrahydro-5,7,7,10,10-pentamethyl-5H-benzo[e]naphtho[2,3-b][1,4]diazepin-13-yl)benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

202110

 
r & d systems
202110
10mg Each for $188.60

202150

 
r & d systems
202150
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 438.56
Quantity 10mg
CAS 155877-83-1
Recommended Storage Store at +4°C
Chemical Name or Material LE 135
Synonym 4-(7,8,9,10-Tetrahydro-5,7,7,10,10-pentamethyl-5H-benzo[e]naphtho[2,3-b][1,4]diazepin-13-yl)benzoic acid
Assay Percent Range >97%
Molecular Formula C29H30N2O2
Safety and Handling

Store at +4°C