Please login to your online account to display your discounted pricing

LDN 57444, Tocris™

Ubiquitin C-terminal hydrolase-L1 (UCH-L1) inhibitor

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material LDN 57444
Assay Percent Range >98%
Molecular Formula C17H11Cl3N2O3
Formula Weight 397.64
Synonym 5-Chloro-1-[(2, 5-dichlorophenyl)methyl]-1H-indole-2, 3-dione 3-(O-acetyloxime)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

399850

 
r & d systems
RDS399850
50mg Each for $697.40

399810

 
r & d systems
RDS399810
10mg Each for $168.15
Description & Specifications

Specifications

Formula Weight 397.64
Quantity 50mg
Chemical Name or Material LDN 57444
Assay Percent Range >98%
Molecular Formula C17H11Cl3N2O3
CAS 668467-91-2
Synonym 5-Chloro-1-[(2, 5-dichlorophenyl)methyl]-1H-indole-2, 3-dione 3-(O-acetyloxime)
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C