missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human Lck, His Tag Recombinant Protein
LCK Recombinant Human Protein
Supplier: Invitrogen™ PV4081
Description
LCK is a SRC-related kinase family expressed in lymphocytes.Specifications
NP_005347 | |
60.6 kDa | |
LCK | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Full Length | |
Signal Transduction |
Kinase Assay | |
1mg | |
SRC Kinase Family | |
Recombinant | |
Storage Buffer: 50 mM Tris, pH 7.5, 100 mM NaCl, 0.05 mM EDTA, 0.05% NP-40, 1 mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.