Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ LCK (p56 lck) Ab-1, Mouse Monoclonal Antibody

Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Thermo Scientific™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results. 

$271.92 - $1,195.98

Specifications

Antigen LCK (p56superscriptlcksuperscript) Ab-1
Clone 3A5
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS164P

 
thermo scientific™ lab vision™
MS164P
200μg/mL 1mL Each for $1,100.51

MS164P0

 
thermo scientific™ lab vision™
MS164P0
200μg/mL 100μL Each for $271.92

MS164P1

 
thermo scientific™ lab vision™
MS164P1
200μg/mL 500μL Each for $697.67

MS164P1ABX

 
thermo scientific™ lab vision™
MS164P1ABX
1mg/mL 100μL Each for $777.22

MS164PABX

 
thermo scientific™ lab vision™
MS164PABX
1mg/mL 200μL Each for $1,195.98
Description & Specifications

Specifications

Antigen LCK (p56superscriptlcksuperscript) Ab-1
Clone 3A5
Applications Immunoprecipitation
Applications Western Blotting
Applications Immunoprecipitation
Applications Western Blotting
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Host Species Murine
Regulatory Status RUO
Research Discipline Cancer and Tumor Biology
Type Primary
Immunogen Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein
Isotype IgG2b kappa

Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Thermo Scientific™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.

Host Species: Mouse

Clone: 3A5

Isotype: IgG2b/κ

Species Reactivity: Human, Mouse and Rat. Others not known.

Epitope: aa 1-225

Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein

Molecular Weight: 56kDa

Positive Control: Jurkat cells

Cellular Localization: Cell membrane

Recommended for:

  • Western Blotting
  • Immunoprecipitation (Denatured verified)
  • Kinase Assay

Note:

USA: RUO; Int'l: RUO