Please login to your online account to display your discounted pricing

Lazabemide hydrochloride, Tocris

Selective MAO-B inhibitor

$149.90 - $632.23

Chemical Identifiers

Chemical Name or Material Lazabemide hydrochloride
Assay Percent Range >98%
Molecular Formula C8H10ClN3O·HCl
Formula Weight 236.1
Synonym N-(2-Aminoethyl)-5-chloro-2-pyridinecarboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

246010

 
r & d systems
RDS246010
10mg Each for $149.90

246050

 
r & d systems
RDS246050
50mg Each for $632.23
Description & Specifications

Specifications

Formula Weight 236.1
Quantity 10mg
Chemical Name or Material Lazabemide hydrochloride
Assay Percent Range >98%
Molecular Formula C8H10ClN3O·HCl
CAS 103878-83-7
Synonym N-(2-Aminoethyl)-5-chloro-2-pyridinecarboxamide hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature