missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Laxco™ LED for MZS32, MZS33, and iLED-30 Series

Laxco™ Wavelength specific LED modules

Manufacturer:  Laxco™ LMSP365

 View more versions of this product

Catalog No. LMSP365



Includes: Single, 365nm LED Module

Description

Description

Laxco™ Wavelength specific LED modules are to be used with Laxco’s ILED-30 Series illuminators and MZS32 & MZS33 Series Microscopes.  Each unit is inspected in the United States by a factory qualified technician under a ridged quality control process to insure performance.

KEY FEATURES:

  • Wavelength specific LED modules
  • 2-Year warranty

Warranty

2 years

Specifications

Specifications

LED
50,000 hours
3W
AMPS-ILED-31; MZS32 Series Stereo Microscopes; MZS33 Series Stereo Microscopes
Single, 365nm LED Module
LED
Stereo Microscopy; Fluorescence Stereo Microscopy; Industrial Applications; Quality Control Applications
LED Module
Documents