Please login to your online account to display your discounted pricing

Lawesson's Reagent, 99%, ACROS Organics™


Quantity 25g
Packaging Glass Bottle in Tin Can

Manufacturer: thermo fisher scientific  210890250

 View more versions of this product

Catalog No. AC210890250

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Lawesson's Reagent
CAS 19172-47-5
Physical Form Powder
Assay ≥98.5% (Titrimetry other)
Assay Percent Range 99%
Molecular Formula C14H14O2P2S4
Formula Weight 404.45
Fieser 10,39; 12,59; 13,38; 15,37; 16,37
Synonym 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane-2,4-disulfide
Gravimetry 98.5% min.
Infrared Spectrum Authentic
Packaging Glass Bottle in Tin Can
Color White to almost white
Melting Point 220° to 226°C
Quantity 25g

Specifications

Chemical Name or Material Lawesson's Reagent
Assay ≥98.5% (Titrimetry other)
Assay Percent Range 99%
Molecular Formula C14H14O2P2S4
Formula Weight 404.45
Synonym 2,4-Bis(4-methoxyphenyl)-1,3-dithia-2,4-diphosphetane-2,4-disulfide