Please login to your online account to display your discounted pricing

Alfa Aesar™ Lavendustin B, 98%

Chemical Identifiers

Chemical Name or Material Lavendustin B
CAS 125697-91-8
Formula Weight 365.39
MDL Number MFCD00133446
Synonym 3-Amino-N,N-bis(2-hydroxybenzyl)salicylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67142MCR

  SDS
alfa aesar™
J67142MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 365.39
Chemical Name or Material Lavendustin B
Assay 98%
Physical Form Solid
MDL Number MFCD00133446
CAS 125697-91-8
Synonym 3-Amino-N,N-bis(2-hydroxybenzyl)salicylic acid
Sensitivity Light Sensitive
Storage Note 1 -30° to -10°C
Color Off-white
Quantity 1mg
Safety and Handling

-30° to -10°C