Please login to your online account to display your discounted pricing

Alfa Aesar™ Lavendustin A


Quantity 1mg

Manufacturer: alfa aesar™  J67494MCR

 View more versions of this product

Catalog No. AAJ67494MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 381.39
Chemical Name or Material Lavendustin A
CAS 125697-92-9
Synonym 5-Amino-N-(2,5-dihydroxybenzyl)-N-(2-hydroxybenzyl)salicylic acid; RG14355
MDL Number MFCD00153822
RTECS Number DG8578950
Sensitivity Air Sensitive
Physical Form Solid
Color Off-white to tan
Quantity 1mg
Storage Note 1 2° to 8°C

Specifications

Chemical Name or Material Lavendustin A
CAS 125697-92-9
Formula Weight 381.39
MDL Number MFCD00153822
Synonym 5-Amino-N-(2,5-dihydroxybenzyl)-N-(2-hydroxybenzyl)salicylic acid; RG14355
Accessories

2° to 8°C