missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Lauryl chloride, 99%, Thermo Scientific Chemicals
$63.58 - $231.25
Chemical Identifiers
CAS | 112-52-7 |
---|---|
Molecular Formula | C12H25Cl |
Molecular Weight (g/mol) | 204.78 |
MDL Number | MFCD00013688 |
InChI Key | YAYNEUUHHLGGAH-UHFFFAOYSA-N |
Synonym | lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk |
PubChem CID | 8192 |
IUPAC Name | 1-chlorododecane |
SMILES | CCCCCCCCCCCCCl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC257102500
|
Thermo Scientific Chemicals
257102500 |
250 mL | Glass bottle |
Each for $63.58
|
|
||||
AC257100010
|
Thermo Scientific Chemicals
257100010 |
1 L | Glass bottle |
Each for $231.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-52-7 | |
204.78 | |
YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
8192 | |
CCCCCCCCCCCCCl |
C12H25Cl | |
MFCD00013688 | |
lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
1-chlorododecane |
Specifications
112-52-7 | |
Colorless | |
260°C | |
Authentic | |
Glass bottle | |
1.4420 to 1.444 | |
MFCD00013688 | |
0.87 | |
Solubility in water: 0.0068g/L (20°C). Other solubilities: miscible with most common organic solvents,slightly miscible with alcohol,soluble in ethanol,benzene,ether,miscible with acetone | |
CCCCCCCCCCCCCl | |
204.78 | |
204.79 | |
Liquid |
-9.3°C | |
0.8700g/mL | |
130°C | |
98.5% min. (GC) | |
C12H25Cl | |
CH3(CH2)11Cl | |
250 mL | |
lauryl chloride, dodecyl chloride, n-dodecyl chloride, dodecane, 1-chloro, laurylchloride, unii-we8o8j0utk, 1-chloro-dodecane, dodecane, chloro, ccris 5810, we8o8j0utk | |
YAYNEUUHHLGGAH-UHFFFAOYSA-N | |
1-chlorododecane | |
8192 | |
99% | |
Lauryl chloride, 99% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Warning
EINECSNumber : 203-981-5
TSCA : TSCA
RUO – Research Use Only