Learn More
Lauroyl chloride, 98%, Thermo Scientific Chemicals
$48.80 - $363.22
Chemical Identifiers
CAS | 112-16-3 |
---|---|
Molecular Formula | C12H23ClO |
Molecular Weight (g/mol) | 218.77 |
MDL Number | MFCD00000740 |
InChI Key | NQGIJDNPUZEBRU-UHFFFAOYSA-N |
Synonym | lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 |
PubChem CID | 8166 |
IUPAC Name | dodecanoyl chloride |
SMILES | CCCCCCCCCCCC(Cl)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC169030500
|
Thermo Scientific Chemicals
169030500 |
50 g | Glass bottle |
Each for $48.80
|
|
||||
AC169032500
|
Thermo Scientific Chemicals
169032500 |
250 g | Glass bottle |
Each for $134.01
|
|
||||
AC169030010
|
Thermo Scientific Chemicals
169030010 |
1 kg | Glass bottle |
Each for $363.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-16-3 | |
218.77 | |
NQGIJDNPUZEBRU-UHFFFAOYSA-N | |
8166 | |
CCCCCCCCCCCC(Cl)=O |
C12H23ClO | |
MFCD00000740 | |
lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 | |
dodecanoyl chloride |
Specifications
112-16-3 | |
0.9200g/mL | |
140°C | |
97.5% min. (GC) | |
C12H23ClO | |
CH3(CH2)10COCl | |
50 g | |
0.92 | |
Solubility in water: reacts. Other solubilities: organic solutions | |
CCCCCCCCCCCC(Cl)=O | |
218.77 | |
218.77 | |
Liquid |
-17.0°C | |
134.0°C to 137.0°C (11.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4440 to 1.4460 | |
MFCD00000740 | |
02, 363 | |
lauroyl chloride, lauroylchloride, n-dodecanoyl chloride, lauric acid chloride, dodecanoic acid, chloride, unii-9lhl10777i, laurel acetyl chlorine, dodecanoylchloride, lauric acid, chloride, pubchem21365 | |
NQGIJDNPUZEBRU-UHFFFAOYSA-N | |
dodecanoyl chloride | |
8166 | |
98% | |
Lauroyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 203-941-7
RUO – Research Use Only