missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Lauronitrile, TCI America™

Manufacturer:  TCI America L0110500ML

 View more versions of this product

Catalog No. L0110500ML



Specifications

Specifications

Lauronitrile
Yellow
2437-25-4
MFCD00001946
VXCUURYYWGCLIH-UHFFFAOYSA-N
dodecanenitrile
17092
Clear Liquid at 20°C
276°C
500mL
C12H23N
lauronitrile, undecyl cyanide, dodecanonitrile, 1-cyanoundecane, n-undecyl cyanide, lauric nitrile, decylacetonitrile, lauric acid nitrile, dodecane nitrile, nitrile 12
CCCCCCCCCCCC#N
181.323
181.32
≥98.0% (GC)

Chemical Identifiers

2437-25-4
181.323
VXCUURYYWGCLIH-UHFFFAOYSA-N
17092
CCCCCCCCCCCC#N
C12H23N
MFCD00001946
lauronitrile, undecyl cyanide, dodecanonitrile, 1-cyanoundecane, n-undecyl cyanide, lauric nitrile, decylacetonitrile, lauric acid nitrile, dodecane nitrile, nitrile 12
dodecanenitrile
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1507

RTECSNumber : JR2600000

TSCA : Yes

Documents