Launch Pro™ Water Rocket Starter Kits
Add water and compressed air.
Manufacturer:  Fisher Scientific 20100*
 View more versions of this product
Catalog No. S94038
Requires :
  • Tap water
  • Various sized plastic soda bottles
  • Compressed air sourse
  • Pressure gauge
  • Materials for constructing fins/rocket bodies
  • Tape
Description
Description
Water rockets are powered by water and compressed air. The compressed air within the bottle expells the water at a high speed to launch the rocket. These kits provide convenient and reliable basic water-rocket launching. Available as Single Stage Kit (S94038) or Multi-Stage Kit (S94037).
  • Mounted launching mechanism
  • Adjustable tripod base folds for compact storage
  • Manual illustrates interdisciplinary experiments combining science, math, and technology
Specifications
Specifications
Hub, 3 legs, fueling and pressurizing hose, 3 screws, 3 washers, 3 wingnuts
5 Cs.
Water rocket starter kit
Documents