Launch Pro™ Water Rocket Starter Kits
Add water and compressed air.
Manufacturer:  Fisher Science Education 20110**
 View more versions of this product
Catalog No. S94037
Requires :
  • Tap water
  • Various sized plastic soda bottles
  • Compressed air sourse
  • Pressure gauge
  • Materials for constructing fins/rocket bodies
  • Tape
Specifications Description & Specifications
Specifications
10 Cs.
Water rocket starter kit
Hub, 3 legs, fueling and pressurizing Hose, launch Pro™ first stage bottle, single stage launch mechanism, multi stage launch mechanism