Please login to your online account to display your discounted pricing
Launch Pro™ Water Rocket Starter Kits
Add water and compressed air.
$128.35 - $175.65
Specifications
Type Water rocket starter kit
View More Specs
Includes:  Base Assembly Kit:
  • Hub
  • Three legs
  • Fueling and pressurizing hose
  • Three each of screws, washers, and wingnuts
Multi-Stage (S94037) also includes:
  • Launch Pro First Stage bottle
  • Launch Pro Second Stage assembly wrench
Launcher Components Kit:
  • Launch Pro Launcher Core assembly
  • Launch Pro Release Cup
  • Launch Pro Bolt
  • Safety clip
  • Two o-rings
  • Two tethers
Multi-Stage (94037) contains two Launcher Components Kits
Requires: 
  • Tap water
  • Various sized plastic soda bottles
  • Compressed air sourse
  • Pressure gauge
  • Materials for constructing fins/rocket bodies
  • Tape
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    
S94037
fisher science education
20110**
Hub, 3 legs, fueling and pressurizing Hose, launch Pro™ first stage bottle, single stage launch mechanism, multi stage launch mechanism 10 Cs. Each for $175.65
Case of 5 Each for $878.30
S94038
fisher science education
20100*
Hub, 3 legs, fueling and pressurizing hose, 3 screws, 3 washers, 3 wingnuts 5 Cs. Each for $128.35
Case of 10 Each for $1,283.15
Description & Specifications
Specifications
Type Water rocket starter kit
Water rockets are powered by water and compressed air. The compressed air within the bottle expells the water at a high speed to launch the rocket. These kits provide convenient and reliable basic water-rocket launching. Available as Single Stage Kit (S94038) or Multi-Stage Kit (S94037).
  • Mounted launching mechanism
  • Adjustable tripod base folds for compact storage
  • Manual illustrates interdisciplinary experiments combining science, math, and technology